Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • APOE Antibodies

NeoBiotechnologies

Apolipoprotein E/APOE Monoclonal Antibody (APOE/3671)

View all (58) APOE antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Apolipoprotein E/APOE Monoclonal Antibody (APOE/3671)

  • Antibody Testing Data (4)
Apolipoprotein E/APOE Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Apolipoprotein E/APOE Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Apolipoprotein E/APOE Antibody (348-MSM1-P0) in IHC (P)

Formalin-fixed, paraffin-embedded human adrenal gland stained with Apolipoprotein E (APOE) Mouse Monoclonal Antibody (APOE/3671). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Apolipoprotein E/APOE Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Apolipoprotein E/APOE Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Apolipoprotein E/APOE Antibody in Peptide array (ARRAY)
Apolipoprotein E/APOE Antibody in SDS-PAGE (SDS-PAGE)
Apolipoprotein E/APOE Monoclonal Antibody (APOE/3671)

Product Details

348-MSM1-P0

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

APOE/3671

Immunogen

Human recombinant APOE protein fragment (around aa19-119).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13. 2.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Apo-E; ApoE4; Apolipoprotein; Apolipoprotein E; apolipoprotein E3; apolipoprotein E4; Apoprotein; B2G1; BG; MGC1571

View more View less

Gene Aliases: AD2; APO-E; APOE; LDLCQ5; LPG

View more View less

UniProt ID: (Human) P02649

View more View less

Entrez Gene ID: (Human) 348

View more View less

Function(s)
beta-amyloid binding lipid transporter activity protein binding phospholipid binding heparin binding lipid binding cholesterol binding antioxidant activity cholesterol transporter activity identical protein binding protein homodimerization activity metal chelating activity tau protein binding low-density lipoprotein particle receptor binding phosphatidylcholine-sterol O-acyltransferase activator activity very-low-density lipoprotein particle receptor binding lipoprotein particle binding
Process(es)
response to reactive oxygen species retinoid metabolic process negative regulation of endothelial cell proliferation response to dietary excess triglyceride metabolic process cholesterol catabolic process cellular calcium ion homeostasis receptor-mediated endocytosis cytoskeleton organization G-protein coupled receptor signaling pathway nitric oxide mediated signal transduction synaptic transmission, cholinergic cholesterol metabolic process regulation of gene expression negative regulation of platelet activation positive regulation of cholesterol esterification positive regulation of cholesterol efflux long-chain fatty acid transport protein import virion assembly triglyceride catabolic process cGMP-mediated signaling negative regulation of blood coagulation regulation of axon extension positive regulation of cGMP biosynthetic process neuron projection regeneration regulation of Cdc42 protein signal transduction positive regulation of low-density lipoprotein particle receptor catabolic process cholesterol efflux phospholipid efflux very-low-density lipoprotein particle remodeling low-density lipoprotein particle remodeling high-density lipoprotein particle remodeling high-density lipoprotein particle assembly chylomicron remnant clearance high-density lipoprotein particle clearance very-low-density lipoprotein particle clearance lipoprotein metabolic process lipoprotein biosynthetic process lipoprotein catabolic process vasodilation cholesterol homeostasis negative regulation of MAP kinase activity negative regulation of neuron apoptotic process negative regulation of blood vessel endothelial cell migration reverse cholesterol transport positive regulation by host of viral process negative regulation of cholesterol biosynthetic process positive regulation of lipid biosynthetic process intracellular transport regulation of neuronal synaptic plasticity artery morphogenesis negative regulation of inflammatory response positive regulation of nitric-oxide synthase activity positive regulation of membrane protein ectodomain proteolysis maintenance of location in cell fatty acid homeostasis positive regulation of dendritic spine development negative regulation of canonical Wnt signaling pathway AMPA glutamate receptor clustering NMDA glutamate receptor clustering cellular oxidant detoxification regulation of beta-amyloid clearance negative regulation of neuron death positive regulation of postsynaptic membrane organization negative regulation of presynaptic membrane organization negative regulation of beta-amyloid formation positive regulation of dendritic spine maintenance positive regulation of phospholipid efflux positive regulation of lipid transport across blood brain barrier
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0