NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody is stable for 24 months.
Positive Control: Human liver, kidney or spleen. Cellular Localization: Secreted.
Specificity Comments: Lipids, such as phospholipids, triacylglycerols and cholesterol, are weakly soluble in aqueous solution and therefore are transported by circulation as components of lipoproteins. Lipoproteins are globular particles that consist of a non-polar core of triacylglycerols and cholesteryl esters surrounded by phospholipid, cholesterol and an amphiphilic coating of protein, known as apolipoproteins (apo). These complexes allow the dissolution and shuttling of their non-polar lipid components. At least nine different apolipoproteins are distributed in significant amounts in different human lipoproteins. Apolipoprotein D (apoD) is a member of the lipocalin superfamily of transporter proteins that bind small hydrophobic molecules, including arachidonic acid (AA). The ability of apoD to bind AA implicates it in pathways associated with membrane phospholipid signal transduction and metabolism. apoD expression has been shown to correlate both with cell cycle arrest and with prognosis in several types of malignancy, including central nervous system astrocytomas and medulloblastomas.
Apolipoprotein D (APOD) is a component of high density lipoprotein that has no marked similarity to other apolipoprotein sequences. It has a high degree of homology to plasma retinol-binding protein and other members of the alpha 2 microglobulin protein superfamily of carrier proteins, also known as lipocalins. This glycoprotein is closely associated with the enzyme lecithin:cholesterol acyltransferase - an enzyme involved in lipoprotein metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apo-D; APO-D protein; Apolipoprotein D
Gene Aliases: APOD
UniProt ID: (Human) P05090
Entrez Gene ID: (Human) 347
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support