Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Angiotensinogen Antibodies

Bioss

Angiotensinogen Polyclonal Antibody, Alexa Fluor™ 647

View all (42) Angiotensinogen antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Angiotensinogen Polyclonal Antibody, Alexa Fluor™ 647

Angiotensinogen Polyclonal Antibody, Alexa Fluor™ 647

Product Details

BS-20778R-A647

Applications
Tested Dilution
Publications

Western Blot (WB)

1:300-5,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunohistochemistry (Frozen) (IHC (F))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from mouse Angiotensinogen
View immunogen

Conjugate

Alexa Fluor™ 647 Alexa Fluor™ 647 Alexa Fluor™ 647

Excitation/Emission Max

650/671 nm View spectra spectra

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

TBS, pH 7.4, with 50% glycerol, 1% BSA

Contains

0.03% ProClin 300

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

The protein encoded by AGT, pre-angiotensinogen or angiotensinogen precursor, is expressed in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. The protein is involved in maintaining blood pressure and in the pathogenesis of essential hypertension and preeclampsia. Mutations in this gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in this gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: alpha-1 antiproteinase, antitrypsin; angiotensin I; angiotensin II; angiotensin ll; Angiotensinogen; angiotensinogen (PAT); angiotensinogen (serpin peptidase inhibitor, clade A, member 8); pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; Serpin A8; serpin peptidase inhibitor, clade A, member 8

View more View less

Gene Aliases: AGT; AI265500; Ang; AngI; AngII; ANHU; ANRT; Aogen; PAT; SERPINA8

View more View less

UniProt ID: (Human) P01019, (Rat) P01015

View more View less

Entrez Gene ID: (Human) 183, (Rat) 24179, (Mouse) 11606

View more View less

Function(s)
serine-type endopeptidase inhibitor activity hormone activity protein binding growth factor activity type 1 angiotensin receptor binding type 2 angiotensin receptor binding sodium channel regulator activity angiotensin receptor binding
Process(es)
regulation of cell growth positive regulation of cytokine production kidney development blood vessel remodeling angiotensin maturation regulation of blood volume by renin-angiotensin renin-angiotensin regulation of aldosterone production regulation of renal output by angiotensin regulation of heart rate regulation of blood vessel size by renin-angiotensin renal system process angiotensin-mediated drinking behavior positive regulation of extracellular matrix constituent secretion cellular sodium ion homeostasis cell surface receptor signaling pathway G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger phospholipase C-activating G-protein coupled receptor signaling pathway activation of phospholipase C activity nitric oxide mediated signal transduction cell-cell signaling female pregnancy aging regulation of blood pressure positive regulation of activation of JAK2 kinase activity positive regulation of endothelial cell migration positive regulation of cardiac muscle hypertrophy positive regulation of macrophage derived foam cell differentiation positive regulation of cholesterol esterification negative regulation of endopeptidase activity regulation of norepinephrine secretion positive regulation of phosphatidylinositol 3-kinase signaling artery smooth muscle contraction response to muscle activity involved in regulation of muscle adaptation negative regulation of angiogenesis regulation of vasoconstriction negative regulation of cell growth positive regulation of cellular protein metabolic process positive regulation of superoxide anion generation positive regulation of NAD(P)H oxidase activity negative regulation of tissue remodeling low-density lipoprotein particle remodeling catenin import into nucleus regulation of renal sodium excretion positive regulation of renal sodium excretion regulation of cell proliferation vasodilation positive regulation of nitric oxide biosynthetic process positive regulation of epidermal growth factor receptor signaling pathway positive regulation of blood pressure positive regulation of transcription, DNA-templated positive regulation of vasodilation fibroblast proliferation positive regulation of fibroblast proliferation regulation of long-term neuronal synaptic plasticity smooth muscle cell proliferation cytokine secretion positive regulation of inflammatory response positive regulation of peptidyl-tyrosine phosphorylation positive regulation of NF-kappaB transcription factor activity negative regulation of neurotrophin TRK receptor signaling pathway stress-activated MAPK cascade regulation of calcium ion transport regulation of transmission of nerve impulse cell growth involved in cardiac muscle cell development positive regulation of protein tyrosine kinase activity ERK1 and ERK2 cascade uterine smooth muscle contraction cellular response to mechanical stimulus positive regulation of branching involved in ureteric bud morphogenesis regulation of extracellular matrix assembly positive regulation of gap junction assembly regulation of cardiac conduction positive regulation of reactive oxygen species metabolic process positive regulation of extrinsic apoptotic signaling pathway ovarian follicle rupture blood vessel development branching involved in ureteric bud morphogenesis regulation of systemic arterial blood pressure by circulatory renin-angiotensin angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure renal response to blood flow involved in circulatory renin-angiotensin regulation of systemic arterial blood pressure brain renin-angiotensin system cell-matrix adhesion activation of NF-kappaB-inducing kinase activity excretion establishment of blood-nerve barrier positive regulation of cell proliferation negative regulation of cell proliferation response to cold response to salt stress regulation of gene expression positive regulation of gene expression extracellular matrix organization peristalsis positive regulation of peptidyl-serine phosphorylation positive regulation of multicellular organism growth vasoconstriction hormone metabolic process drinking behavior regulation of apoptotic process positive regulation of catalytic activity positive regulation of MAPK cascade negative regulation of neuron apoptotic process positive regulation of fatty acid biosynthetic process positive regulation of organ growth astrocyte activation regulation of inflammatory response smooth muscle cell differentiation positive regulation of protein kinase C activity positive regulation of membrane hyperpolarization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0