Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Acetylcholinesterase Antibodies

Rockland

Acetyl Cholinesterase Polyclonal Antibody

View all (20) Acetylcholinesterase antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Acetyl Cholinesterase Polyclonal Antibody

Acetyl Cholinesterase Polyclonal Antibody

Product Details

200-101-232-0100

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:5,000
-

ELISA (ELISA)

1:5,000-1:20,000
-

Immunoprecipitation (IP)

1:500-1:5,000
-
Product Specifications

Species Reactivity

Bovine

Host/Isotype

Goat / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Acetyl Cholinesterase [Bovine Erythrocytes]
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Ion-exchange chromatography

Storage buffer

0.02M potassium phosphate, pH 7.2, with 0.15M NaCl

Contains

0.01% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Wet ice

Product Specific Information

Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.

Anti-ACETYL CHOLINESTERASE is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum as well as purified and partially purified Acetyl Cholinesterase [Bovine Erythrocytes]. Cross reactivity against Acetyl Cholinesterase from other sources is unknown.

Target Information

Acetylcholinesterase hydrolyzes the neurotransmitter, acetylcholine at neuromuscular junctions and brain cholinergic synapses, and thus terminates signal transmission. The Protein is also found on the red blood cell membranes, where it constitutes the Yt blood group antigen. Acetylcholinesterase exists in multiple molecular forms which possess similar catalytic properties, but differ in their oligomeric assembly and mode of cell attachment to the cell surface. The major form of acetylcholinesterase found in brain, muscle and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, or lipid-containing structural subunits.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Acetylcholinesterase; acetylcholinesterase (Yt blood group); AChE; EC 3.1.1.7; OTTHUMP00000211349; OTTHUMP00000211356

View more View less

Gene Aliases: ACHE; BOS_23015

View more View less

UniProt ID: (Bovine) P23795

View more View less

Entrez Gene ID: (Bovine) 540446

View more View less

Function(s)
beta-amyloid binding acetylcholinesterase activity cholinesterase activity protein binding carboxylic ester hydrolase activity
Process(es)
synaptic transmission, cholinergic synapse assembly choline metabolic process neurotransmitter catabolic process regulation of dendrite morphogenesis regulation of axonogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-c7d89468c-sbmwt:80/100.66.128.142:80.
git-commit: 4db89d68e0ee702f860d53b64def1eb94897fade
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0