Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ATP5J Antibodies

Proteintech

ATP5J Polyclonal Antibody

2 References
View all (8) ATP5J antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ATP5J Polyclonal Antibody

  • Antibody Testing Data (7)
ATP5J Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
ATP5J Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 7

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

ATP5J Antibody (14114-1-AP) in IHC (P)

Immunohistochemistry of paraffin-embedded human osteosarcoma using 14114-1-AP (ATP5J antibody) at dilution of 1:100 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

ATP5J Antibody in Immunohistochemistry (Paraffin) (IHC (P))
ATP5J Antibody in Immunohistochemistry (Paraffin) (IHC (P))
ATP5J Antibody in Western Blot (WB)
ATP5J Antibody in Western Blot (WB)
ATP5J Antibody in Western Blot (WB)
ATP5J Antibody in Western Blot (WB)
ATP5J Antibody in Immunoprecipitation (IP)
ATP5J Polyclonal Antibody

Product Details

14114-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
View 2 publications 2 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:20-1:200
-

Immunoprecipitation (IP)

0.5-4.0 µg
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

ATP5J Fusion Protein Ag5263 (1-108 aa encoded by BC066310)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.4 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: MILQRLFRF SSVIRSAVSV HLRRNIGVTA VAFNKELDPI QKLFVDKIRE YKSKRQTSGG PVDASSEYQQ ELERELFKLK QMFGNADMNT FHTFKFEDPK FEVIEKPQA (1-108 aa encoded by BC066310 )

Target Information

Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. It is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, F0, which comprises the proton channel. The F1 complex consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled in a ratio of 3 alpha, 3 beta, and a single representative of the other 3. The F0 seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the F6 subunit of the F0 complex, required for F1 and F0 interactions. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: ATP synthase peripheral stalk subunit F6; ATP synthase peripheral stalk subunit F6, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F6; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6; ATP synthase-coupling factor 6, mitochondrial; ATPase subunit F6; C ATP5; coupling factor 6; mitochondrial ATP synthase, coupling factor 6; mitochondrial ATP synthase, subunit F6; mitochondrial ATPase coupling factor 6; proliferation-inducing protein 36

View more View less

Gene Aliases: ATP5; ATP5A; ATP5J; ATP5PF; ATPM; CF6; F6

View more View less

UniProt ID: (Human) P18859, (Mouse) P97450, (Rat) P21571

View more View less

Entrez Gene ID: (Human) 522, (Mouse) 11957, (Rat) 94271

View more View less

Function(s)
transporter activity hydrogen ion transmembrane transporter activity ATPase activity transmembrane transporter activity
Process(es)
ATP biosynthetic process substantia nigra development mitochondrial ATP synthesis coupled proton transport ATP synthesis coupled proton transport ATP metabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0