Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ATP Synthase gamma Antibodies

Proteintech

ATP5C1 Monoclonal Antibody (1B5A2)

4 References
View all (14) ATP Synthase gamma antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ATP5C1 Monoclonal Antibody (1B5A2)

  • Antibody Testing Data (5)
ATP5C1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
ATP5C1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

ATP5C1 Antibody (60284-1-IG) in ICC/IF

Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using (Product # 60284-1-IG) (ATP5C1 antibody), at dilution of 1:100 and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

ATP5C1 Antibody in Immunocytochemistry (ICC/IF)
ATP5C1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
ATP5C1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
ATP5C1 Antibody in Western Blot (WB)
ATP5C1 Antibody in Western Blot (WB)
ATP5C1 Monoclonal Antibody (1B5A2)

Product Details

60284-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
View 4 publications 4 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:100-1:400
-

Immunocytochemistry (ICC/IF)

1:100
-
Product Specifications

Species Reactivity

Human

Published species

Human

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

1B5A2

Immunogen

ATP5C1 Fusion Protein Ag1343 (1-298 aa encoded by BC000931)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite 594
  • Request custom conjugation

Form

Liquid

Concentration

0.77 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: MFSRAGVAG LSAWTLQPQW IQVRNMATLK DITRRLKSIK NIQKITKSMK MVAAAKYARA ERELKPARIY GLGSLALYEK ADIKGPEDKK KHLLIGVSSD RGLCGAIHSS IAKQMKSEVA TLTAAGKEVM LVGIGDKIRG ILYRTHSDQF LVAFKEVGRK PPTFGDASVI ALELLNSGYE FDEGSIIFNK FRSVISYKTE EKPIFSLNTV ASADSMSIYD DIDADVLQNY QEYNLANIIY YSLKESTTSE QSARMTAMDN ASKNASEMID KLTLTFNRTR QAVITKELIE IISGAAALD (1-298 aa encoded by BC000931 )

Target Information

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: ATP synthase F(1) complex subunit gamma, mitochondrial; ATP synthase F1 subunit gamma; ATP synthase gamma chain, mitochondrial; F-ATPase gamma subunit; mitochondrial ATP synthase, gamma subunit 1

View more View less

Gene Aliases: ATP5C; ATP5C1; ATP5CL1; ATP5F1C

View more View less

UniProt ID: (Human) P36542

View more View less

Entrez Gene ID: (Human) 509

View more View less

Function(s)
ATPase activity transmembrane transporter activity poly(A) RNA binding proton-transporting ATP synthase activity, rotational mechanism proton-transporting ATPase activity, rotational mechanism
Process(es)
oxidative phosphorylation ATP biosynthetic process mitochondrial ATP synthesis coupled proton transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0