Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Apolipoprotein A5 Antibodies

OriGene

APOA5 Monoclonal Antibody (OTI1E3), TrueMAB™

View all (29) Apolipoprotein A5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite APOA5 Monoclonal Antibody (OTI1E3), TrueMAB™

  • Antibody Testing Data (2)
APOA5 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
APOA5 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

APOA5 Antibody (TA507094) in WB

Western blot analysis of extracts (35 µg) from 9 different cell lines by using anti-APOA5 monoclonal antibody (HepG2: human; HeLa: human; SVT2: mouse; A549: human; COS7: monkey; Jurkat: human; MDCK: canine; PC12: rat; MCF7: human). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

APOA5 Antibody in Western Blot (WB)
APOA5 Antibody in Western Blot (WB)
APOA5 Monoclonal Antibody (OTI1E3), TrueMAB™

Product Details

TA507094

Applications
Tested Dilution
Publications

Western Blot (WB)

1:4,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI1E3

Immunogen

Full length human recombinant protein of human APOA5 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but several evidences from vitro studies indicate that this apolipoprotein involves in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Apo-AV; Apolipoprotein A-V; apolipoprotein A-V precursor variant 3; Apolipoprotein A5; apolipoprotein AV; FLJ97995; MGC126836; MGC126838; Regeneration-associated protein 3

View more View less

Gene Aliases: APOA5; APOAV; RAP3; UNQ411/PRO773

View more View less

UniProt ID: (Human) Q6Q788

View more View less

Entrez Gene ID: (Human) 116519

View more View less

Function(s)
phospholipid binding enzyme activator activity heparin binding lipid binding cholesterol binding cholesterol transporter activity enzyme binding phosphatidylcholine binding lipase binding low-density lipoprotein particle receptor binding phosphatidylcholine-sterol O-acyltransferase activator activity lipase activator activity lipoprotein lipase activator activity lipoprotein particle receptor binding
Process(es)
triglyceride metabolic process cholesterol biosynthetic process lipid transport positive regulation of cholesterol esterification positive regulation of triglyceride catabolic process positive regulation of very-low-density lipoprotein particle remodeling triglyceride catabolic process regulation of intestinal cholesterol absorption neuron projection regeneration cholesterol efflux phospholipid efflux triglyceride-rich lipoprotein particle remodeling high-density lipoprotein particle assembly lipoprotein metabolic process tissue regeneration cholesterol homeostasis reverse cholesterol transport positive regulation of fatty acid biosynthetic process phosphatidylcholine metabolic process positive regulation of receptor-mediated endocytosis positive regulation of lipid catabolic process positive regulation of lipoprotein lipase activity acylglycerol homeostasis triglyceride homeostasis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0