Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • APLP2 Antibodies

Invitrogen

APLP2 Recombinant Rabbit Monoclonal Antibody (23GB1595)

Advanced Verification
View all (12) APLP2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite APLP2 Recombinant Rabbit Monoclonal Antibody (23GB1595)

  • Antibody Testing Data (3)
  • Advanced Verification (1)
APLP2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
APLP2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

APLP2 Antibody (MA5-53664) in ICC/IF

Immunocytochemistry analysis of APLP2 in HepG2 cells. Nuclei were stained blue with DAPI; APLP2 recombinant monoclonal antibody (Product # MA5-53664) with a dilution of 1:1,000 was stained magenta with Alexa Fluor 647. Images were taken using Leica stellaris 5. Protein abundance based on laser Intensity and smart gain: High. Scale bar: 20 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

APLP2 Antibody in Immunocytochemistry (ICC/IF)
APLP2 Antibody in Western Blot (WB)
APLP2 Antibody in Flow Cytometry (Flow)
APLP2 Antibody
APLP2 Recombinant Rabbit Monoclonal Antibody (23GB1595)

Product Details

MA5-53664

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:5,000
-

Immunocytochemistry (ICC/IF)

1:1,000
-

Flow Cytometry (Flow)

1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293F

Class

Recombinant Monoclonal

Type

Antibody

Clone

23GB1595

Immunogen

A synthesized peptide derived from human APLP2 (221-280AA).
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3248135

Target Information

APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5'-GTCACATG-3' (CDEI box).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Amyloid beta (A4) precursor-like protein 2; Amyloid beta precursor like protein 2; amyloid precursor protein homolog HSD-2; Amyloid protein homolog; Amyloid-like protein 2; APLP-2; APPH; CDEBP; CDEI box-binding protein; Sperm membrane protein YWK-II; testicular tissue protein Li 23

View more View less

Gene Aliases: APLP-2; APLP2; APPH; APPL2; CDEBP

View more View less

UniProt ID: (Human) Q06481

View more View less

Entrez Gene ID: (Human) 334

View more View less

Function(s)
DNA binding serine-type endopeptidase inhibitor activity protein binding heparin binding identical protein binding transition metal ion binding
Process(es)
platelet degranulation G-protein coupled receptor signaling pathway negative regulation of endopeptidase activity
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0