Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ANGPTL7 Antibodies

Invitrogen

ANGPTL7 Polyclonal Antibody

View all (14) ANGPTL7 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ANGPTL7 Polyclonal Antibody

  • Antibody Testing Data (4)
ANGPTL7 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
ANGPTL7 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

ANGPTL7 Antibody (PA5-106604) in ICC/IF

Immunocytochemistry analysis of ANGPTL7 in Hela cells. The samples were fixed with PFA and permeabilized in 0.1% Triton X-100, then blocked in 10% serum for 45 minutes at 25°C. Samples were incubated with ANGPTL7 polyclonal antibody (Product # PA5-106604) and mouse anti-beta tubulin antibody for 1 hour at 37°C. An AlexaFluor594 conjugated goat anti-rabbit IgG(H+L) (Red) and an AlexaFluor488 conjugated goat anti-mouse IgG(H+L) (Green) were used as the secondary antibodies. The nuclear counter stain is DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

ANGPTL7 Antibody in Immunocytochemistry (ICC/IF)
ANGPTL7 Antibody in Western Blot (WB)
ANGPTL7 Antibody in Western Blot (WB)
ANGPTL7 Antibody in Western Blot (WB)
ANGPTL7 Polyclonal Antibody

Product Details

PA5-106604

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:1,000
-

Immunocytochemistry (ICC/IF)

1:100-1:500
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

A synthesized peptide derived from human ANGPTL7(Accession O43827), corresponding to amino acid residues E312-P346.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2854272

Product Specific Information

Antibody detects endogenous levels of total ANGPTL7.

Target Information

Angptl7 (angiopoietin-like 7), alternately known as cornea-derived transcript 6 protein (CDT6) or AngX, is a 346 amino acid secreted protein that is expressed specifically in the stromal layer of cornea. Angptl7 forms disulfide-linked homotetramers via coiled-coil interactions and contains one fibrinogen C-terminal domain. Overexpressed in patients with glaucoma, Angptl7 serves as a potential therapeutic target for glaucoma and has been found to increase collagen expression. The gene encoding Angptl7 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: angiopoietin-like 7; Angiopoietin-like factor; angiopoietin-like factor (CDT6); Angiopoietin-like protein 7; Angiopoietin-related protein 7; Cornea-derived transcript 6 protein

View more View less

Gene Aliases: ANGPTL7; AngX; CDT6; dJ647M16.1; UNQ313/PRO356

View more View less

UniProt ID: (Human) O43827, (Mouse) Q8R1Q3

View more View less

Entrez Gene ID: (Human) 10218, (Mouse) 654812

View more View less

Process(es)
response to oxidative stress
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0