Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • AMH Antibodies

Invitrogen

AMH Recombinant Rabbit Monoclonal Antibody (501)

View all (41) AMH antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite AMH Recombinant Rabbit Monoclonal Antibody (501)

  • Antibody Testing Data (1)
AMH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
AMH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

AMH Antibody (MA5-52353) in IHC (P)

Immunohistochemistry analysis of AMH in formalin-fixed paraffin embedded human testis sections. Samples were incubated with AMH monoclonal antibody (Product # MA5-52353) using a dilution of 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

AMH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
AMH Recombinant Rabbit Monoclonal Antibody (501)

Product Details

MA5-52353

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

501

Immunogen

Recombinant Human AMH Protein (Leu25-Arg560)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

100 µg/mL

Purification

Protein A

Storage buffer

PBS

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3248830

Target Information

Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: AMH; Anti-Muellerian hormone; MIS; Muellerian-inhibiting factor; Muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance

View more View less

Gene Aliases: AMH; MIF; MIS

View more View less

UniProt ID: (Human) P03971

View more View less

Entrez Gene ID: (Human) 268

View more View less

Function(s)
receptor binding transforming growth factor beta receptor binding hormone activity growth factor activity
Process(es)
preantral ovarian follicle growth urogenital system development Mullerian duct regression cell-cell signaling gonadal mesoderm development sex determination sex differentiation aging positive regulation of gene expression response to organic cyclic compound response to drug positive regulation of NF-kappaB transcription factor activity negative regulation of ovarian follicle development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0