Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD5L Antibodies

Invitrogen

AIM Recombinant Rabbit Monoclonal Antibody (11H48L16)

Advanced Verification
View all (19) CD5L antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite AIM Recombinant Rabbit Monoclonal Antibody (11H48L16)

  • Antibody Testing Data (1)
  • Advanced Verification (1)
AIM Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
AIM Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

AIM Antibody (703558) in ICC/IF

For immunofluorescence analysis, RAW 264.7 cells were fixed and permeabilized for detection of endogenous AIM using Anti-AIM Recombinant Rabbit Monoclonal Antibody (Product # 703558, 1:100 dilution) and labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product # A27034, 1:2000). Nuclei (blue) were stained using ProLong™ Diamond Antifade Mountant with DAPI (Product # P36962) and cytoskeletal F-actin (red) staining using Rhodamine Phalloidin (Product # R415, 1:300) Panel a-d) shows r... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

AIM Antibody in Immunocytochemistry (ICC/IF)
AIM Antibody
AIM Recombinant Rabbit Monoclonal Antibody (11H48L16)

Product Details

703558

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1:100
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Rabbit / IgG

Expression System

Expi293

Class

Recombinant Monoclonal

Type

Antibody

Clone

11H48L16

Immunogen

Peptides corresponding to Mouse AIM [1) aa42-aa54 2) aa271-aa283]
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

0.09% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Wet ice

RRID

AB_2762393

Product Specific Information

This antibody is predicted to react with Monkey, Rat.

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Target Information

CD5L, also known as CD5 antigen-like, is a protein that is expressed on the surface of various immune cells, including T cells, B cells, and natural killer cells. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including autoimmune disorders, cancer, and infectious diseases. CD5L is known to modulate T cell activation and cytokine production, and has been shown to have anti-inflammatory effects.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: AIM; AIM/Spalpha; apoptosis inhibitor 6; Apoptosis inhibitor expressed by macrophages; APOPTOSIS INHIBITOR OF MACROPHAGES; Apoptosis inhibitory 6; CD5 antigen like (scavenger receptor cysteine rich family); CD5 antigen-like; CT-2; Highly similar to ANTIGEN WC1.1 [Bos taurus]; mAIM; Pdp 1/6; SCAVENGER RECEPTOR CYSTEINE-RICH FAMILY; SP-alpha

View more View less

Gene Aliases: 1/6; AAC-11; AI047839; Aim; Api6; Cd5l; CT2; Pdp; Sp-alpha

View more View less

UniProt ID: (Mouse) Q9QWK4

View more View less

Entrez Gene ID: (Mouse) 11801

View more View less

Function(s)
scavenger receptor activity
Process(es)
apoptotic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0