Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ADAR1 Antibodies

Invitrogen

ADAR1 Recombinant Rabbit Monoclonal Antibody (7H12)

View all (22) ADAR1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ADAR1 Recombinant Rabbit Monoclonal Antibody (7H12)

  • Antibody Testing Data (1)
ADAR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
ADAR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

ADAR1 Antibody (MA5-49891) in IHC (P)

Immunohistochemistry analysis with a ADAR1 monoclonal antibody (Product # MA5-49891) using a dilution of 1:100 and staining in paraffin-embedded human brain tissue performed on a Leica BondTM system. After dewaxing and hydration, antigen retrieval was mediated by high pressure in a citrate buffer (pH 6.0). Section was blocked with 10% normal goat serum 30 min at RT. Then primary antibody (1% BSA) was incuba... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

ADAR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

Product Details

MA5-49891

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

7H12

Immunogen

A synthesized peptide derived from human ADAR1

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.25 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3092807

Target Information

Adenosine Deaminase RNA Specific (ADAR) catalyzes the hydrolytic deamination of adenosine to inosine in double-stranded RNA (dsRNA) referred to as A-to-I RNA editing.This may affect gene expression and function in a number of ways that include mRNA translation by changing codons and hence the amino acid sequence of proteins; pre-mRNA splicing by altering splice site recognition sequences; RNA stability by changing sequences involved in nuclease recognition; genetic stability in the case of RNA virus genomes by changing sequences during viral RNA replication; and RNA structure-dependent activities such as microRNA production or targeting or protein-RNA interactions. ADAR can edit both viral and cellular RNAs and can edit RNAs at multiple sites (hyper-editing) or at specific sites (site-specific editing). Its cellular RNA substrates include: bladder cancer-associated protein (BLCAP), neurotransmitter receptors for glutamate (GRIA2) and serotonin (HTR2C) and GABA receptor (GABRA3). Site-specific RNA editing of transcripts encoding these proteins results in amino acid substitutions which consequently alters their functional activities. Exhibits low-level editing at the GRIA2 Q/R site, but edits efficiently at the R/G site and HOTSPOT1. Its viral RNA substrates include: hepatitis C virus (HCV), vesicular stomatitis virus (VSV), measles virus (MV), hepatitis delta virus (HDV), and human immunodeficiency virus type 1 (HIV-1). Exhibits either a proviral (HDV, MV, VSV and HIV-1) or an antiviral effect (HCV) and this can be editing-dependent (HDV and HCV), editing-independent (VSV and MV) or both (HIV-1). Impairs HCV replication via RNA editing at multiple sites. Enhances the replication of MV, VSV and HIV-1 through an editing-independent mechanism via suppression of EIF2AK2/PKR activation and function. Stimulates both the release and infectivity of HIV-1 viral particles by an editing-dependent mechanism where it associates with viral RNAs and edits adenosines in the 5'UTR and the Rev and Tat coding sequence. Can enhance viral replication of HDV via A-to-I editing at a site designated as amber/W, thereby changing an UAG amber stop codon to an UIG tryptophan (W) codon that permits synthesis of the large delta antigen (L-HDAg) which has a key role in the assembly of viral particles. However, high levels of ADAR1 inhibit HDV replication. (Uniprot)

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 136 kDa double-stranded RNA-binding protein; ADA; Adenosine Deaminase; adenosine deaminase acting on RNA 1-A; adenosine deaminase, RNA-specific; Double-stranded RNA-specific adenosine deaminase; DRADA; dsRNA adenosine deaminase; dsRNA adeonosine deaminase; IFI-4; interferon-induced protein 4; Interferon-inducible protein 4; K88DSRBP; p136

View more View less

Gene Aliases: ADAR; ADAR1; AGS6; DRADA; DSH; DSRAD; G1P1; IFI-4; IFI4; K88DSRBP; P136

View more View less

UniProt ID: (Human) P55265

View more View less

Entrez Gene ID: (Human) 103

View more View less

Function(s)
DNA binding double-stranded RNA adenosine deaminase activity protein binding poly(A) RNA binding metal ion binding
Process(es)
osteoblast differentiation in utero embryonic development hematopoietic progenitor cell differentiation somatic diversification of immune receptors via somatic mutation adenosine to inosine editing mRNA processing response to virus base conversion or substitution editing erythrocyte differentiation pre-miRNA processing miRNA loading onto RISC involved in gene silencing by miRNA response to interferon-alpha negative regulation of apoptotic process negative regulation of protein kinase activity by regulation of protein phosphorylation positive regulation of viral genome replication negative regulation of viral genome replication innate immune response defense response to virus definitive hemopoiesis type I interferon signaling pathway negative regulation of type I interferon-mediated signaling pathway hematopoietic stem cell homeostasis cellular response to virus negative regulation of RNA interference
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0